Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17C Mouse anti-Human, APC, Clone: 177114, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523567

Catalog No. PIMA523567


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit human IL-17A, B, D, E or F is observed.

Cytokine that plays a crucial role in innate immunity of the epithelium, including to intestinal bacterial pathogens, in an autocrine manner. Stimulates the production of antibacterial peptides and proinflammatory molecules for host defense by signaling through the NF-kappa-B and MAPK pathways. Acts synergically with IL22 in inducing the expression of antibacterial peptides, including S100A8, S100A9, REG3A and REG3G. Synergy is also observed with TNF and IL1B in inducing DEFB2 from keratinocytes. Depending on the type of insult, may have both protective and pathogenic properties, either by maintaining epithelial homeostasis after an inflammatory challenge or by promoting inflammatory phenotype. Enhanced IL17C/IL17RE signaling may also lead to greater susceptibility to autoimmune diseases.
Specifications

Specifications

IL17C
Monoclonal
APC
IL17C
Liquid
IL17C
E. coli-derived recombit human IL-17C His19-Val197
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
177114
Buffered saline solution with BSA and 0.09% sodium azide
Q9P0M4
CX2, IL-17C, IL-21, cytokine CX2, interleukin-17C
Mouse
IgG2A
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
27189
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit