Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17RB Rat anti-Mouse, Clone: 752101, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524360

Catalog No. PIMA524360


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human (rh) IL-17 RB, rhIL-17 R, or recombit mouse IL-17 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are IL-17B, IL-17C, and IL-17E. Like IL-17, IL-17B, C, and E are able to induce proinflammatory responses but don ft.t bind to the IL-17 receptor suggesting additional IL-17R related receptors might exist. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in kidney and liver and at moderate level in small intestine, colon, and brain.
Specifications

Specifications

IL17RB
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9JIP3
CRL4, EVI27, IL17BR, IL17RH1, MGC5245
Rat
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
50905
Flow Cytometry
752101
Unconjugated
Il17rb
Lyophilized
IL17RB
Mouse myeloma cell line NS0-derived recombit mouse IL-17RB Arg18-Gly286
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit