Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17RD Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526636

Catalog No. PIPA526636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, rat and Xenopus based on sequence homology.

Fibroblast growth factors (FGFs; see MIM 603726) are secreted proteins involved in cellular proliferation, migration, differentiation, and survival. FGF activity is negatively regulated by members of the  ft.sprouty ft. family (e.g., SPRY1, MIM 602465). The SEF protein is a modulator of FGF signaling.
Specifications

Specifications

IL17RD
Polyclonal
PBS with 0.09% sodium azide
Q8NFM7
IL-17RD, IL17RLM, SEF, IL-17 receptor D, IL17Rhom, hSef, interleukin-17 receptor D, interleukin-17 receptor-like protein, sef homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54756
Western Blot
Unconjugated
IL17RD
Liquid
IL17RD
KLH conjugated synthetic peptide between 217-245 amino acids from the N-terminal region of human IL17RD
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit