Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL1F5, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177873

Catalog No. NBP177873


Add to cart

Description

Description

IL-36Ra/IL-1F5 Polyclonal antibody specifically detects IL-36Ra/IL-1F5 in Bovine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IL1F5
LYOPH
Western Blot 1 ug/ml, ELISA 1:8000
Purified
IL36RN
This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein raised in yeast, corresponding to the 155 amino acids of the mature bovine IL-1F5 protein.
Protein A purified
RUO
Primary
26525
Reconstitute with 100 ul deionized water.
ELISA, Western Blot
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1HY1, IL1HY1IL-1-related protein 3, IL-1L1, IL1L1MGC29840, IL-1RP3, IL1RP3IL-1ra homolog 1, interleukin 1 family, member 5 (delta), interleukin 1, delta, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1
Rabbit
IgG
0.1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for bovine IL-1F5 protein. A BLAST analysis was used to suggest cross-reactivity with IL-1F5 from dog and mouse sources based on 94% identity with the immunizing sequence. Partial reactivity is expected against horse based on 91% homology, and to human, chimpanzee, macaque, and rat based on 90% homology with the immunizing sequence.
Bovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only