Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1RN, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant IL1RN.

Manufacturer:  Abnova Corporation H00003557A01

Catalog No. 89-003-009


Add to cart

Description

Description

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1, alpha (IL1A) and interleukin 1, beta (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. This gene and five other closely related cytokine genes form a gene cluster spanning approximately 400kb on chromosome 2. A polymorphism of this gene is reported to be associated with increased risk of osteoporotic fractures and gastric cancer. Four alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: MALETICRPSGRKSSKMQAFRIWDVNQKTFYLRNNQLVAGYLQGPNVNLEEKIDVVPIEPHALFLGIHGGKMCLSCVKSGDETRLQLEAVNITDLSENRKQDKRFAFIRSDSGPTTSFESAACPGWFLCTAMEADQPVSLTNMPDEGVMVTKFYFQEDE
Specifications

Specifications

IL1RN
Unconjugated
50% glycerol
BC009745
ICIL-1RA|IL-1ra3|IL1F3|IL1RA|IRAP|MGC10430
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a full-length recombinant IL1RN.
IL1RN
Liquid
IL1RN
IL1RN (AAH09745, 1 a.a. ~ 159 a.a) full-length recombinant protein with GST tag.
RUO
Primary
3557
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit