Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL24 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525789

Catalog No. PIPA525789


Add to cart

Description

Description

Interleukin 24 (IL-24) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in maligt gliomas in a p53-independent manner.
Specifications

Specifications

IL24
Polyclonal
Unconjugated
IL24
Liquid
IL24
KLH conjugated synthetic peptide between 139-168 amino acids from the C-terminal region of human IL24
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q13007
C49A, FISP, IL10B, MDA7, MOB5, ST16, IL-4-induced secreted protein, interleukin-24, melanocyte-associated Mda-7, melanoma differentiation-associated gene 7 protein, suppression of tumorigenicity 16 (melanoma differentiation)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11009
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit