Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RG Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526461

Catalog No. PIPA526461


Add to cart

Description

Description

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
Specifications

Specifications

IL2RG
Polyclonal
PBS with 0.09% sodium azide
P31785
IL2RG, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1, CD132 antigen, IL-2 receptor subunit gamma, IL-2R subunit gamma, cytokine receptor common subunit gamma, gamma (c), gammaC, interleukin-2 receptor subunit gamma, IL2R gamma
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3561
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
IL2RG
Liquid
IL2RG
KLH conjugated synthetic peptide between 76-101 amino acids from the N-terminal region of human IL2RG
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit