Learn More
IL8, Mouse, Clone: 4E3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant IL8.
Supplier: Abnova Corporation H00003576M18
Description
The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. [provided by RefSeq
Sequence: EGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENSSpecifications
IL8 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC013615 | |
IL8 | |
IL8 (AAH13615, 21 a.a. ∼ 99 a.a) full length recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunoprecipitation, Western Blot | |
4E3 | |
Mouse monoclonal antibody raised against a full length recombinant IL8. | |
IL8 | |
CXCL8/GCP-1/GCP1/LECT/LUCT/LYNAP/MDNCF/MONAP/NAF/NAP-1/NAP1 | |
Mouse | |
Affinity chromatography | |
RUO | |
3576 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.