Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin 8, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human IL8 protein.

Manufacturer:  Abnova Corporation H00003576B01

Catalog No. 89-005-628


Add to cart

Description

Description

The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. [provided by RefSeq

Sequence: MTSKLAVALLAAFLISAALCEGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS
Specifications

Specifications

interleukin 8
Unconjugated
No additive
BC013615
CXCL8|GCP-1|GCP1|LECT|LUCT|LYNAP|MDNCF|MONAP|NAF|NAP-1|NAP1
Mouse
50 uL
Angiogenesis
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human IL8 protein.
IL8
Serum
IL8
IL8 (AAH13615, 1 a.a. ~ 99 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3576
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit