Learn More
integrin-linked kinase-associated serine/threonine phosphatase 2C (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ILKAP.
Supplier: Abnova Corporation H00080895A01
Description
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway. [provided by RefSeq
Sequence: IGDGQYKRCGVTSVPDIRRCQLTPNDRFILLACDGLFKVFTPEEAVNFILSCLEDEKIQTREGKSAADARYEAACNRLANKAVQRGSADNVTVMVVRIGHSpecifications
integrin-linked kinase-associated serine/threonine phosphatase 2C | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ILKAP. | |
ILKAP | |
DKFZp434J2031/FLJ10181/MGC4846/PP2C-DELTA | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_030768 | |
ILKAP | |
ILKAP (NP_110395, 293 a.a. ∼ 392 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
80895 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.