Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ILT7/CD85g/LILRA4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215013
Description
ILT7/CD85g/LILRA4 Polyclonal specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Western Blot.Specifications
ILT7/CD85g/LILRA4 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
CD85 antigen-like family member G, CD85g, CD85g antigen, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597 | |
Rabbit | |
55 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:1000-1:10000 | |
P59901 | |
LILRA4 | |
Recombinant protein encompassing a sequence within the center region of human LILRA4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
23547 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only