Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ILVBL, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178433AF405

Catalog No. NBP178433W


Add to cart

Description

Description

ILVBL Polyclonal antibody specifically detects ILVBL in Human samples. It is validated for Western Blot.
Specifications

Specifications

ILVBL
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
ILVBL
A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Western Blot
Western Blot
A1L0T0
acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535
Rabbit
IgG
0.1 ml
Epigenetics
Primary
10994
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only