Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILVBL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | ILVBL |
---|---|
Concentration | 0.20 mg/mL |
Dilution | Western Blot 1:1000, ICC/IF (Negative) |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP178433SS
|
Novus Biologicals
NBP178433SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP178433
|
Novus Biologicals
NBP178433 |
0.1 mL |
Each for $409.50
|
|
|||||
Description
ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
ILVBL | |
Western Blot 1:1000, ICC/IF (Negative) | |
Polyclonal | |
Rabbit | |
Epigenetics | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 | |
ILVBL | |
IgG | |
Affinity Purified |
0.20 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Human | |
A1L0T0 | |
10994 | |
A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title