Learn More
Imipramine Mouse anti-Chemical, Clone: IM12-379.2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110724
Catalog No. 01-671-954
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10724 detects Imipramine. MA1-10724 has been successfully used in ELISA applications. The MA1-10724 immunogen is Imipramine, conjugated to KLH.
Imipramine Base is a synthetic tricyclic derivative, antidepressant Imipramine enhances monoamine neurotransmission in certain areas of the brain. It also induces sedation through histamine 1 receptor blockage; hypotension through beta-adrenergic blockage; and diverse parasympatholytic effects. Imipramine has less sedative effect than other members of its therapeutic family. It is used in major depression, dysthymia, bipolar depression, attention-deficit disorders, agoraphobia, and panic disorders. [National Center for Biotechnology Information. PubChem Compound Database; CID=3696]
Specifications
Specifications
Imipramine
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Antideprin; Deprenil; Deprimin; Deprinol; Depsonil; Dynaprin; Eupramin; Imipramil; Irmin; Janimine; Melipramin; Surplix; Tofranil; C19H24N2
Imipramine, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
IM12-379.2
Unconjugated
Liquid
Mouse
IgG2b
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical