Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IMP Dehydrogenase 2/IMPDH2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128478

Catalog No. NBP128478

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IMP Dehydrogenase 2/IMPDH2
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
NP_000875.2
EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMPD 2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2
Goat
IgG
0.1 mg
Lipid and Metabolism
Primary
3615
Human
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
IMPDH2
Peptide with sequence KEEEHDCFLEEI corresponding to internal region according to NP_000875.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only