Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

inositol(myo)-1(or 4)-monophosphatase 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human IMPA2 protein.

Manufacturer:  Abnova Corporation H00003613B01

Catalog No. 89-005-639


Add to cart

Description

Description

Sequence: MKPSGEDQAALAAGPWEECFQAAVQLALRAGQIIRKALTEEKRVSTKTSAADLVTETDHLVEDLIISELRERFPSHRFIAEEAAASGAKCVLTHSPTWIIDPIDGTCNFVHRFPTVAVSIGFAVRQELEFGVIYHCTEERLYTGRRGRGAFCNGQRLRVSGETDLSKALVLTEIGPKRDPATLKLFLSNMERLLHAKAHGVRVIGSSTLALCHLASGAADAYYQFGLHCWDLAAATVIIREAGGIVIDTSGGPLDLMACRVVAASTREMAMLIAQALQTINYGRDDEK
Specifications

Specifications

inositol(myo)-1(or 4)-monophosphatase 2
Unconjugated
No additive
BC017176.1
IMPA2
IMPA2 (AAH17176.1, 1 a.a. ~ 288 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3613
Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human IMPA2 protein.
IMPA2
Serum
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit