Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPDH2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527519

Catalog No. PIPA527519


Add to cart

Description

Description

PA5-27519 targets IMPDH2 in IF and WB applications and shows reactivity with Human and mouse samples. The PA5-27519 immunogen is recombit fragment corresponding to a region within amino acids 276 and 506 of Human IMPDH2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5 ft.-monophosphate into xanthine-5 ft.-monophosphate, which is then converted into guanosine-5 ft.-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in maligt transformation.
Specifications

Specifications

IMPDH2
Polyclonal
Unconjugated
IMPDH2
Liquid
IMPDH2
Recombit fragment corresponding to a region within amino acids 276 and 506 of Human IMPDH2
Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P12268, P24547
IMPD2, IMPDH-II, IMP (inosine monophosphate) dehydrogenase 2, IMP oxireductase 2, IMPD 2, IMPDH 2, inosine 5′ phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5′-monophosphate dehydrogenase 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23918, 3615
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit