Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Importin 11, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233209

Catalog No. NBP233209


Add to cart

Description

Description

Importin 11 Polyclonal antibody specifically detects Importin 11 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Importin 11
0.5 mg/mL
Western Blot 0.5-2 ug/ml
Q9UI26
imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51194
Human, Mouse, Rat, Bovine, Canine, Primate
Western Blot
Unconjugated
PBS containing 0.2% gelatin. Does not contain BSA. with No Preservative
Affinity Purified
IPO11
A portion of amino acids 1-50 of human Importin-11 was used as the immunogen (NP_001128251).
112 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Bovine, Chimpanzee, Dog, Human, Mouse, Rat, Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Xenopus (94%).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only