Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Importin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | Importin-9 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056489T
|
Novus Biologicals
NB10056489SS |
0.025 mg |
Each for $204.00
|
|
|||||
NB10056489
|
Novus Biologicals
NB10056489 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
Importin-9 Polyclonal specifically detects Importin-9 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Importin-9 | |
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Primate | |
Q96P70 | |
55705 | |
A portion of amino acids 950-1000 of human Importin-9 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 | |
IPO9 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b and CRA_c), chimp, rhesus monkey, mouse (isoforms 1 and 2 and Importin-9a and Importin-9b), rat and bovine and 82% homologous in chicken. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title