Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Importin beta-1 Mouse anti-Human, Clone: 3H14, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051530
Description
Use Anti-Importin beta-1 Antibody, clone 3H14 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect Importin beta-1 also known as Karyopherin subunit beta-1, Nuclear factor P97.Specifications
Importin beta-1 | |
Monoclonal | |
Unconjugated | |
NP_002256.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
3H14 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
KPNB1, IMB1, IPO1, IPOB, Impnb, Importin-90, NTF97 | |
Recombinant protein corresponding to human Importin beta-1. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only