Learn More
Importin subunit alpha-1 Mouse anti-Human, Clone: 8G5, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051526
Catalog No. 05-152-6MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-Importin subunit alpha-1 Antibody, clone 8G5 is a mouse monoclonal antibody for detection of Importin subunit alpha-1 also known as karyopherin alpha 1, importin alpha 5, importin-alpha-S1 and has been validated in western blotting.
Specifications
Specifications
Importin subunit alpha-1
Monoclonal
Unconjugated
NP_002255.3
Mouse
Protein G purfied
Primary
2-8°C for one year
IgG1 κ
Western Blot
8G5
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
RCH2, NPI-1, SRP1, SRP1-beta, IPOA5, KPNA1
Recombinant protein corresponding to human Importin subunit alpha-1.
100 μg
Human
Purified