Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INO80B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524764

Catalog No. PIPA524764


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

IN80B induces growth and cell cycle arrests at the G1 phase of the cell cycle.
Specifications

Specifications

INO80B
Polyclonal
PBS with 0.09% sodium azide
Q9C086
HMGA1L4, HMGIYL4, IES2, PAP-1BP, PAPA-1, ZNHIT4, hIes2, IES2 homolog, PAP (Pim-1 associated protein) associated protein 1, PAP-1 binding protein, PAP-1-associated protein 1, high mobility group AT-hook 1-like 4, zinc finger HIT domain-containing protein 4, zinc finger, HIT type 4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83444
Flow Cytometry, Western Blot
Unconjugated
INO80B
Liquid
INO80B
KLH conjugated synthetic peptide between 146-174 amino acids from the central region of human IN80B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit