Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Internexin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513283
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
INA is a class-IV neuronal intermediate filament that is able to self-assemble. It is involved in the morphogenesis of neurons. It may form an independent structural network without the involvement of other neurofilaments or it may cooperate with NF-L to form the filamentous backbone to which NF-M and NF-H attach to form the cross-bridges.Specifications
alpha Internexin | |
Polyclonal | |
INA | |
Alpha-internexin; Alpha-Inx; 66 kDa neurofilament protein; Neurofilament-66; NF-66; NEF5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9118 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q16352 | |
INA | |
KLH conjugated synthetic peptide between 290-319 amino acids from the central region of human INA (alpha internexin) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title