Learn More
MilliporeSigma™ anti-Influenza A, Clone: H16-L10-4R5, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB8800/DEL
Encompass
Catalog No. 50-175-846
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
ELISA, Flow Cytometry, Immunofluorescence, Western Blotting
Specifications
Specifications
Influenza A
Unconjugated
100 μL
Viral And Bacterial
Primary
Semi-purified
Monoclonal
Mouse
RUO
Specific to the Influenza A nuclear protein
Human
IgG2a
Product Suggestions
Product Suggestions