Learn More
Influenza A M1, HRP, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA127357
Catalog No. PA127357
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Influenza A virus matrix 1, M1, is a critical protein required for assembly and budding. Hemagglutinin associates with M1 via its cytoplasmic tail and transmembrane domain. The M2 and nucleoprotein proteins are critical in the replication cycle of influenza viruses. The M2 channel protein is an essential component of the viral envelope because of its ability to form a highly selective, pH-regulated, proton-conducting channel. The M2 channel allows protons to enter the interior of the virus and acidification weakens the interaction of the M1 protein with the ribonuclear core.
PA1-27357 detects Influenza A Virus M1 Protein from virus samples. PA1-27357 will not react with Influenza B, Adenovirus, Respiratory syncytial virus, or Parainfluenza viruses (1-3).PA1-27357 has been successfully used in ELISA procedures.The PA1-27357 immunogen is Influenza A strain Phillipines (H3N2).Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Specifications
Specifications
Influenza A M1
Polyclonal
HRP
Purified
Goat
IgG
100μL
Store at -20°C
Primary
Virus
ELISA, Immunocytochemistry, Immunofluorescence
1.5mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Influenza Matrix protein M1, Influenza M1
Influenza A strain Phillipines (H3N2).
IgG fraction
RUO
Antibody
Polyclonal