Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVNS1ABP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124418

Catalog No. PA124418


Add to cart

Description

Description

PA1-24418 detects Influenza Virus NS1A Binding Protein in Human samples. PA1-24418 has been successfully used in Western Blot and Immunohistochemistry (paraffin) procedures. PA1-24418 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human IVNS1ABP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

IVNS1ABP is a novel human protein that interacts with the influenza A virus. Nonstructural NS1 protein is relocalized in the nuclei of infected cells.
Specifications

Specifications

IVNS1ABP
Polyclonal
Unconjugated
IVNS1ABP
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10625
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y6Y0
IVNS1ABP
Polyclonal antibody from rabbits immunized with a synthetic peptide region of human IVNS1ABP
Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit