Learn More
anti-Influenza A, Nucleoprotein, Clone: A3, Millipore™ (Chemicon™)
Manufacturer:  Millipore Sigma MAB8258/DEL
 View more versions of this product
Catalog No. 50-175-758
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Influenza A, nucleoprotein
Human
Murine
IgG
RUO
Specific for the influenza A nucleoprotein. Stronger reactivity is observed with N2/N3-type Flu A strains.
Monoclonal
Unlabeled
Purified
Influenza A
100μg
Viral And Bacterial
Primary
Human