Learn More
Influenza A H1N1, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA127308
Catalog No. PA127308
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
PA1-27308 detects Influenza A Virus from virus samples. Does not react with Influenza B, RSV, Parainfluenza 1-3 or Adenovirus. Does not react with HEp-2 cells.PA1-27308 has been successfully used in immunohistochemistry (paraffin), immunofluorescence and ELISA procedures.The PA1-27308 immunogen is Influenza A strain USSR (H1N1).Store at 4ºC short term. For long term storage, store at -20ºC, avoiding freeze/thaw cycles.
Specifications
Specifications
Influenza A H1N1
Polyclonal
Unconjugated
Purified
Goat
IgG
100μL
Store at -20°C
Primary
Virus
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
Influenza H1N1, H1N1
Influenza A strain USSR (H1N1)
IgG fraction
RUO
Antibody
Polyclonal