Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Virions Goat anti-Virus, HRP, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173045

Catalog No. PIPA173045


Add to cart

Description

Description

The PA1-73045 antibody reacts with Influenza A virions H1N1. It does not react with Influenza B, RSV, Para 1-3 or adeno viruses. PA1-73045 has been successfully used in ELISA applications. The PA1-73045 antibody was raised against Influenza A, strain USSR (H1N1). NOTE: sodium azide is a potent inhibitor of peroxidase.

Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifications

Specifications

Influenza A H1N1 Virions
Polyclonal
HRP
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Influenza H1N1; H1N1; H1N1 virions
Influenza A, strain USSR (H1N1).
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit