Learn More
MilliporeSigma™ anti-Influenza B Blend, Clone: B2 and B4 Blend, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB8661/DEL
Encompass
Catalog No. 50-175-811
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
Immunofluorescence
Specifications
Specifications
Influenza B Blend
Unconjugated
Influenza Blend.
All subtypes of Influenza B. Blend of two clones that recognize the nucleoprotein antigen.
Human
IgG
Monoclonal
Mouse
100 μL
Primary
Ascites
Product Suggestions
Product Suggestions