Learn More
Influenza B Virions, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA130853
Catalog No. PA130853
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
PA1-30853 detects Influenza B from virus samples.PA1-30853 has been successfully used in ELISA, immunofluorescence, Western blot applications.The PA1-30853 immunogen is: Influenza B virions, Yamagata strain
Specifications
Specifications
Influenza B Virions
Polyclonal
Unconjugated
Purified
Goat
IgG1
1mL
Store at -20°C
Primary
Virus
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
Influenza Virions
Influenza B virions, Yamagata strain
IgG fraction
RUO
Antibody
Polyclonal