Learn More
ING1 Mouse anti-Human, Clone: 3E7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519208
Catalog No. PIMA519208
Add to cart
Description
Description
Peptide Sequence: DAKYQEILKE LDECYERFSR ETDGAQKRRM LHCVQRALIR SQELGDEKIQ IVSQMVELVE NRTRQVDSHV ELFEAQQELG DTAGNSGKAG ADRPKGEAAA
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
Specifications
ING1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9UK53
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth protein 1, tumor suppressor ING1
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3621
ELISA, Western Blot
3E7
Unconjugated
ING1
Liquid
ING1
ING1 (NP_937862.1, 41 a.a. ∽ 140 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human