Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527716

Catalog No. PIPA527716


Add to cart

Description

Description

PA5-27716 targets ING1 in WB applications and shows reactivity with Human samples. The PA5-27716 immunogen is synthetic peptide corresponding to a region within amino acids 271 and 367 of Human ING1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

ING1
Polyclonal
Unconjugated
ING1
Liquid
ING1
Synthetic peptide corresponding to a region within amino acids 271 and 367 of Human ING1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q9UK53
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth protein 1, tumor suppressor ING1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3621
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit