Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

inhibitor of growth family, member 2 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ING2 protein.

Manufacturer:  Abnova Corporation H00003622B02

Catalog No. 89-005-644



Description

Description

This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis. [provided by RefSeq

Sequence: MLGQQQQQLYSSAALLTGERSRLLTCYVQDYLECVESLPHDMQRNVSVLRELDNKYQETLKEIDDVYEKYKKEDDLNQKKRLQQLLQRALINSQELGDEKIQIVTQMLELVENRARQMELHSQCFQDPAESERASDKAKMDSSQPERSSRRPRRQRTSESRDLCHMANGIEDCDDQPPKEKKSKSAKKKKRSKAKQEREASPVEFAIDPNEPTYCLCNQVSYGEMIGCDNEQCPIEWFHFSCVSLTYKPKGKWYCPKCRGDNEKTMDKSTEKTKKDRRSR
Specifications

Specifications

inhibitor of growth family, member 2
Unconjugated
No additive
NM_001564.2
ING1L|p33ING2
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3622
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ING2 protein.
ING2
Serum
ING2
ING2 (NP_001555.1, 1 a.a. ~ 280 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit