Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Inhibin beta A, Polyclonal, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196131

Catalog No. NBP196131

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Inhibin beta A
1.8 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:25 - 1:100, Immunohistochemistry-Paraffin 1:25 - 1:100
Affinity Purified
INHBA
INHBA fusion protein ag16700
47 kDa
15 ml
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3) and 50% Gycerol with 0.02% Sodium Azide
Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3624
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only