Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iNOS Mouse anti-Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610600

 View more versions of this product

Catalog No. BDB610600


Add to cart

Description

Description

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In macrophages and other cell types, NOS (iNOS or macNOS) activity increases following exposure to cytokines (IFN-γ, TNF-α, and IL-1) and microbial products (lipopolysaccharide (LPS)). iNOS isactivated independently of Ca2+/calmodulin and its level of expression is tightly controlled by several transcription factors, including NFκB. Data indicates that TGF-β affects translation of iNOS mRNA and decreases iNOS protein stability. Normally undetectable in brain tissue, iNOS mRNA has been observed in CNS tissues of animals under experimental pathologic conditions. iNOS and nNOS share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

iNOS
2
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
NOS Type II
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.