Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPP5A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528158

Catalog No. PIPA528158


Add to cart

Description

Description

PA5-28158 targets INPP5A in WB applications and shows reactivity with Human samples. The PA5-28158 immunogen is recombit fragment corresponding to a region within amino acids 86 and 352 of Human INPP5A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a membrane-associated type I inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase. InsP3 5-phosphatases hydrolyze Ins(1,4,5)P3, which mobilizes intracellular calcium and acts as a second messenger mediating cell responses to various stimulation.
Specifications

Specifications

INPP5A
Polyclonal
Unconjugated
INPP5A
Liquid
INPP5A
Recombit fragment corresponding to a region within amino acids 86 and 352 of Human INPP5A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.74 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q14642
5PTASE, 43 kDa inositol polyphosphate 5-phophatase, CTCL tumor antigen HD-CL-02, InsP3 5-phosphatase, inositol trisphosphate-5-phosphatase, 40kD, type I inositol-1,4,5-trisphosphate 5-phophatase, type I inositol-1,4,5-trisphosphate 5-phosphatase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3632
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit