Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPP5B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523992

Catalog No. PIPA523992

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). INPP5B encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications

Specifications

INPP5B
Polyclonal
PBS with 0.09% sodium azide
P32019
5PTase, 75 kDa inositol polyphosphate-5-phosphatase, phosphoinositide 5-phosphatase, type II inositol-1,4,5-trisphosphate 5-phosphatase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3633
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
INPP5B
Liquid
INPP5B
KLH conjugated synthetic peptide between 950-979 amino acids from the C-terminal region of human INPP5B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit