Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPP5F Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521562

Catalog No. PIPA521562


Add to cart

Description

Description

PA5-21562 targets INPP5F in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-21562 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 593 and 904 (Q9Y2H2) of INPP5F. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

INPP5F
Polyclonal
Unconjugated
INPP5F
Liquid
INPP5F
Recombit protein fragment contain a sequence corresponding to a region within amino acids 593 and 904 (Q9Y2H2) of INPP5F
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.79 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9Y2H2, Q8CDA1
MSTP007, MSTPO47, SAC2, hSAC2, Sac domain-containing inositol phosphatase 2, phosphatidylinositide phosphatase SAC2, sac domain-containing phosphoinositide 5-phosphatase 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
101490, 22876
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit