Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526707

Catalog No. PIPA526707

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization.
Specifications

Specifications

INSC
Polyclonal
PBS with 0.09% sodium azide
Q1MX18, Q3HNM7
protein inscuteable homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
233752, 387755
Immunohistochemistry, Western Blot
Unconjugated
INSC
Liquid
INSC
KLH conjugated synthetic peptide between 409-437 amino acids from the C-terminal region of human INSC
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit