Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INSIG-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB11055244SS
Description
INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
INSIG-1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
Rabbit | |
30 kDa | |
0.025 mL | |
Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
3638 | |
Human, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.02 mg/mL | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
O15503 | |
INSIG1 | |
A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503. | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for Insig1 but not Insig2. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction