Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Mouse anti-Bovine, Human, Porcine, Clone: D4B8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA110516

Catalog No. 01-671-532



Description

Description

MA1-10516 detects Insulin from human, bovine, and porcine samples. MA1-10516 has been successfully used in ELISA and immunohistochemistry (frozen) applications. The MA1-10516 immunogen is purified human Insulin.

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
Specifications

Specifications

Insulin
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P01317, P01308, P01315
Ins
Mouse
IgG1
200 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
280829, 3630, 397415
ELISA, Immunohistochemistry (Frozen)
D4B8
Unconjugated
INS
Liquid
INS
Purified human Insulin.
Protein A
RUO
Antibody
Monoclonal
Bovine, Human, Porcine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit