Learn More
MilliporeSigma™ anti-Integrin β1, a.a. 82-87 (CD29), Clone: JB1A (J10), ™ (Chemicon™)
Manufacturer: MilliporeSigma™ MAB1965/DEL

Description
The primary function of the integrin family is to mediate cell-cell and cell-matrix adhesion interaction. CD29 is ubiquitous, with broad tissue distribution, but is not expressed on erythrocytes and is expressed only weakly on granulocytes. Integrins play an important role in cell adhesion and migration, and their normal function is critical in the induction and maintenance of cell differentiation. Loss or downregulation of CD29 has been proposed to be one of the general pathways through which carcinoma cells may acquire a more invasive and differentiated phenotype. Furthermore, loss or downregulation of integrin occurs relatively frequently in poorly differentiated colorectal and breast carcinomas as determined by immunohistochemistry.
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
CD29/Integrin β1, a.a. 82-87 | |
Unconjugated | |
Purified β 1 integrin and Jurkat cells | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Cell Structure | |
Human | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.