Learn More
Integrin α1 Mouse anti-Human, Clone: 5E8D9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 05246
Catalog No. 05-246-MI
Add to Cart
Description
Description
Anti-Integrin α1 Antibody, clone 5E8D9 is an antibody against Integrin α1 for use in immunocytochemistry, NEUT.
Specifications
Specifications
Integrin α1
Monoclonal
Unconjugated
NM_181501
Mouse
Protein A chromatography
Primary
−20°C for 24 months.
IgG2a
Immunocytochemistry, Neutralization
5E8D9
0.1M Tris-glycine, pH 7.4, 0.15M NaCl
ITGA1, CD49a, VLA1, VLA-1
Purified human integrinα1
200 μg
Human
Purified
Recommendations
Recommendations