Learn More
Integrin α4 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB1919
Catalog No. AB1919MI
Description
Description
This Anti-Integrin α4 Antibody, PhosphoSerine 988 is validated for use in ELISA, immunoprecipitation, western blotting for the detection of Integrin α4.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin α4
Polyclonal
Liquid rabbit serum containing no preservative.
Rabbit
100 μL
NM_000885.4
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ITGA4; MGC90518; VLA-4; IA4; CD49D; CD49d
Synthetic peptide corresponding to the cytoplasmic tail of human integrin alpha4 (a.a. 986-995) containing a phosphorylated serine at position 988. The phosphopeptide was conjugated to KLH using gluteraldehyde.
Primary
Human
Serum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only