Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2 Rat anti-Mouse, PE, Clone: 235033, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523537

Catalog No. PIMA523537


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit mouse Integrin alpha 3, alpha 4, alpha 5, alpha E, or alpha V is observed. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

This gene product belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane glycoproteins composed of a distinct alpha chain and a common beta chain. They are found on a wide variety of cell types including, T cells, fibroblasts and platelets. Integrins are involved in cell adhesion and also participate in cell-surface mediated signalling.
Specifications

Specifications

ITGA2
Monoclonal
PE
Itga2
Liquid
ITGA2
Mouse myeloma cell line NS0-derived recombit mouse Integrin alpha 2/CD49b Tyr27-Thr1129
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
235033
Buffered saline solution with BSA and <0.1% sodium azide
Q62469
BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2
Rat
IgG2B
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
16398
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit