Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5 Mouse anti-Human, APC, Clone: 238307, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523585

Catalog No. PIMA523585


Add to cart

Description

Description

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Specifications

Specifications

ITGA5
Monoclonal
APC
ITGA5
Liquid
ITGA5
Mouse myeloma cell line NS0-derived recombit human Integrin alpha 5/CD49e Phe42-Tyr995
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
238307
Buffered saline solution with BSA and 0.09% sodium azide
P08648
FNRA, CD49e, VLA5A, ITGA5
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3678
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit