Learn More
anti-Integrin alpha M/CD11b, Alexa Fluor 647, Clone: 238446, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB16991R025
 View more versions of this product
Catalog No. FAB16991R02
Add to cart
Specifications
Specifications
CD11b
238446
Flow Cytometry 5 uL/10^6 cells
NP_001139280
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Mouse
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human Integrin alpha M/CD11b in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Supernatant
ITGAM
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha M/CD11b, Phe17-Asn1105, Accession # NP_001139280
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3684
Human