Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-Integrin alpha M/CD11b, PerCP, Clone: 238446, R&D Systems
Manufacturer: R&D Systems FAB16991C100
Specifications
Integrin alpha M/CD11b | |
238446 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Purified | |
Mouse myeloma cell line NS0-derived recombinant human Integrin αM/CD11b, Phe17-Asn1105 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Monoclonal | |
Detects human Integrin αM/CD11b in direct ELISAs. |
Flow Cytometry | |
PerCP | |
NP_001139280 | |
Murine | |
IgG2b | |
100 Tests | |
Primary | |
3684 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title