Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Integrin alphav Clone: 272-17E6
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 407286100UG
Description
Specifically detects Integrin αv Clone: 272-17E6 in Human samples, and it is validated for ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays
The integrins are a family of more than 23 heterodimeric transmembrane proteins that mediate cell-cell adhesions as well as cell-substratum adhesions and signal transduction processes. Integrins are heterodimers consisting of noncovalently associated α and β subunits. Human CD51 is an integrin alpha v chain which can associate with five different beta chains (β1, β3, β5, β6, and β8) to form various adhesion molecules. CD51 is found in endothelial cells, smooth muscle cells, fibroblasts, mast cells, monocytes, osteoclasts, platelets, and melanoma cells.
Specifications
| Integrin αv | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays | |
| 272-17E6 | |
| In PBS, pH 7.6. | |
| human placental αvβ3 integrin | |
| RUO | |
| Recognizes the αv integrin chain and all αv-containing integrins. | |
| −20°C | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction